Assay Details
| Gene Symbol: mnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142022.1 | NP_001135494.1 | 4 - 5 | 1225 | 75 | |
| XM_012956552.2 | XP_012812006.1 | 6 - 7 | 1235 | 75 | ||
| GenBank mRNA | BC167894.1 | - | 4 - 5 | 1225 | 75 | |
More Information
Set Membership: |
|

mnt
Genomic Map