Assay Details
| Gene Symbol: nsd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142024.1 | NP_001135496.1 | 4 - 5 | 1155 | 89 | |
| GenBank mRNA | BC167896.1 | - | 4 - 5 | 1155 | 89 | |
More Information
Set Membership: |
|

nsd3
Genomic Map