Assay Details
| Gene Symbol: rpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142035.1 | NP_001135507.1 | 337 | 68 | ||
| GenBank mRNA | BC167917.1 | - | 337 | 68 | ||
| BC168604.1 | - | 307 | 68 | |||
More Information
Set Membership: |
|

rpl6
Genomic Map