Assay Details
| Gene Symbol: mfsd14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142061.1 | NP_001135533.1 | 9 - 10 | 1100 | 80 | |
| GenBank mRNA | BC121501.1 | - | 10 - 11 | 1622 | 80 | |
| BC167991.1 | - | 9 - 10 | 1100 | 80 | ||
| BC168126.1 | - | 9 - 10 | 1633 | 80 | ||
| BC168629.1 | - | 9 - 10 | 1633 | 80 | ||
More Information
Set Membership: |
|

mfsd14b
Genomic Map