Assay Details
| Gene Symbol: slc9a3r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142062.1 | NP_001135534.1 | 6 - 7 | 1169 | 62 | |
| GenBank mRNA | BC168002.1 | - | 6 - 7 | 1169 | 62 | |
More Information
Set Membership: |
|

slc9a3r2
Genomic Map