Assay Details
| Gene Symbol: lgals8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142086.1 | NP_001135558.1 | 3 - 4 | 195 | 86 | |
| XM_012962795.2 | XP_012818249.1 | 4 - 5 | 86 | |||
| XM_012962797.2 | XP_012818251.1 | 3 - 4 | 86 | |||
| XM_018093808.1 | XP_017949297.1 | 3 - 4 | 86 | |||
| GenBank mRNA | BC168047.1 | - | 3 - 4 | 195 | 86 | |
More Information
Set Membership: |
|

lgals8
Genomic Map