Assay Details
| Gene Symbol: ssbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142090.1 | NP_001135562.1 | 14 - 15 | 980 | 60 | |
| XM_012957436.2 | XP_012812890.1 | 11 - 12 | 1336 | 60 | ||
| XM_012957446.2 | XP_012812900.1 | 13 - 14 | 1126 | 60 | ||
| XM_018091350.1 | XP_017946839.1 | 11 - 12 | 1363 | 60 | ||
| XM_018091351.1 | XP_017946840.1 | 11 - 12 | 1366 | 60 | ||
| XM_018091354.1 | XP_017946843.1 | 10 - 11 | 1281 | 60 | ||
| XM_018091358.1 | XP_017946847.1 | 14 - 15 | 1235 | 60 | ||
| XM_018091362.1 | XP_017946851.1 | 13 - 14 | 1151 | 60 | ||
| GenBank mRNA | BC168053.1 | - | 14 - 15 | 980 | 60 | |
| CT030517.1 | - | 13 - 14 | 874 | 60 | ||
More Information
Set Membership: |
|

ssbp2
Genomic Map