Assay Details
| Gene Symbol: mecp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142091.1 | NP_001135563.1 | 2 - 3 | 391 | 101 | |
| GenBank mRNA | BC168054.1 | - | 2 - 3 | 391 | 101 | |
More Information
Set Membership: |
|

mecp2
Genomic Map