Assay Details
| Gene Symbol: lamtor3-like | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142094.1 | NP_001135566.1 | 6 - 7 | 380 | 89 | |
| GenBank mRNA | BC168057.1 | - | 6 - 7 | 380 | 89 | |
| CU075450.1 | - | 6 - 7 | 372 | 89 | ||
More Information
Set Membership: |
|

lamtor3-like
Genomic Map