Assay Details
| Gene Symbol: lsm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142097.1 | NP_001135569.1 | 4 - 5 | 364 | 103 | |
| GenBank mRNA | BC168072.1 | - | 4 - 5 | 364 | 103 | |
More Information
Set Membership: |
|

lsm5
Genomic Map