Assay Details
Gene Symbol: pkdcc.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142110.1 | NP_001135582.2 | 2 - 3 | 663 | 97 | |
XM_018096176.1 | XP_017951665.1 | 2 - 3 | 1475 | 97 | ||
GenBank mRNA | BC168090.1 | - | 2 - 3 | 663 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
