Assay Details
| Gene Symbol: rbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142134.1 | NP_001135606.1 | 1 - 2 | 232 | 79 | |
| GenBank mRNA | BC168424.1 | - | 1 - 2 | 232 | 79 | |
| CU075387.1 | - | 1 - 2 | 85 | 79 | ||
More Information
Set Membership: |
|

rbp1
Genomic Map