Assay Details
| Gene Symbol: chmp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142136.1 | NP_001135608.1 | 4 - 5 | 351 | 97 | |
| GenBank mRNA | BC168427.1 | - | 4 - 5 | 351 | 97 | |
| CU075373.1 | - | 4 - 5 | 264 | 97 | ||
More Information
Set Membership: |
|

chmp1a
Genomic Map