Assay Details
| Gene Symbol: wt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142153.1 | NP_001135625.1 | 841 | 65 | ||
| GenBank mRNA | BC168476.1 | - | 841 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

wt1| Gene Symbol: wt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142153.1 | NP_001135625.1 | 841 | 65 | ||
| GenBank mRNA | BC168476.1 | - | 841 | 65 | ||
Set Membership: |
|