Assay Details
| Gene Symbol: asphd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142183.1 | NP_001135655.1 | 4 - 5 | 1464 | 99 | |
| XM_012957536.2 | XP_012812990.1 | 4 - 5 | 1654 | 99 | ||
| XM_012957538.2 | XP_012812992.1 | 5 - 6 | 1918 | 99 | ||
| GenBank mRNA | BC168555.1 | - | 4 - 5 | 1464 | 99 | |
More Information
Set Membership: |
|

asphd2
Genomic Map