Assay Details
| Gene Symbol: insyn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142211.1 | NP_001135683.1 | 1 - 2 | 217 | 67 | |
| XM_012966236.2 | XP_012821690.1 | 3 - 4 | 1013 | 67 | ||
| XM_012966237.2 | XP_012821691.1 | 3 - 4 | 1007 | 67 | ||
| GenBank mRNA | BC168617.1 | - | 1 - 2 | 217 | 67 | |
More Information
Set Membership: |
|

insyn1
Genomic Map