Assay Details
| Gene Symbol: aif1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142238.1 | NP_001135710.1 | 3 - 4 | 434 | 73 | |
| XM_012968126.1 | XP_012823580.1 | 4 - 5 | 73 | |||
| GenBank mRNA | BC168836.1 | - | 3 - 4 | 434 | 73 | |
More Information
Set Membership: |
|

aif1l
Genomic Map