Assay Details
| Gene Symbol: epgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142239.2 | NP_001135711.2 | 3 - 4 | 248 | 70 | |
| GenBank mRNA | BC168838.1 | - | 3 - 4 | 70 | ||
| BX703151.1 | - | 3 - 4 | 70 | |||
| CU025080.1 | - | 3 - 4 | 70 | |||
| DR879072.1 | - | 3 - 4 | 224 | 70 | ||
More Information
Set Membership: |
|

epgn
Genomic Map