Assay Details
| Gene Symbol: pms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142901.2 | NP_001136373.2 | 1 - 2 | 155 | 71 | |
| XM_012970376.2 | XP_012825830.1 | 2 - 3 | 250 | 71 | ||
| XM_012970377.2 | XP_012825831.1 | 2 - 3 | 250 | 71 | ||
| GenBank mRNA | BC089718.1 | - | 1 - 2 | 155 | 71 | |
| BC121226.1 | - | 2 - 3 | 216 | 71 | ||
| CR762073.2 | - | 1 - 2 | 168 | 71 | ||
More Information
Set Membership: |
|

pms1
Genomic Map