Assay Details
| Gene Symbol: ccnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142906.1 | NP_001136378.1 | 2 - 3 | 805 | 94 | |
| XM_012962551.2 | XP_012818005.1 | 7 - 8 | 910 | 94 | ||
| GenBank mRNA | BC121713.1 | - | 2 - 3 | 880 | 94 | |
| BC158441.1 | - | 7 - 8 | 758 | 94 | ||
| BC161176.1 | - | 2 - 3 | 805 | 94 | ||
More Information
Set Membership: |
|

ccnl1
Genomic Map