Assay Details
| Gene Symbol: nr5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145741.1 | NP_001139213.1 | 5 - 6 | 1164 | 66 | |
| GenBank mRNA | AB385624.1 | - | 5 - 6 | 1164 | 66 | |
More Information
Set Membership: |
|

nr5a1
Genomic Map