Assay Details
| Gene Symbol: spib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145984.1 | NP_001139456.1 | 3 - 4 | 154 | 73 | |
| GenBank mRNA | FJ644947.1 | - | 3 - 4 | 154 | 73 | |
More Information
Set Membership: |
|

spib
Genomic Map