Assay Details
| Gene Symbol: spib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145984.1 | NP_001139456.1 | 5 - 6 | 517 | 89 | |
| XM_018095307.1 | XP_017950796.1 | 5 - 6 | 608 | 89 | ||
| GenBank mRNA | FJ644947.1 | - | 5 - 6 | 517 | 89 | |
More Information
Set Membership: |
|

spib
Genomic Map