Assay Details
| Gene Symbol: dgcr8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159539.1 | NP_001153011.1 | 12 - 13 | 2241 | 69 | |
| XM_012955055.2 | XP_012810509.1 | 13 - 14 | 2674 | 69 | ||
| XM_012955085.2 | XP_012810539.1 | 13 - 14 | 2304 | 69 | ||
| GenBank mRNA | BC159181.1 | - | 12 - 13 | 1575 | 69 | |
| EU338240.1 | - | 12 - 13 | 2241 | 69 | ||
More Information
Set Membership: |
|

dgcr8
Genomic Map