Assay Details
| Gene Symbol: jagn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001898.1 | NP_001001898.1 | 3 | 452 | 91 | |
| XM_018092984.1 | XP_017948473.1 | 4 | 735 | 91 | ||
| XM_018092985.1 | XP_017948474.1 | 3 | 449 | 91 | ||
| GenBank mRNA | BC067954.1 | - | 3 | 452 | 91 | |
| CR760294.2 | - | 2 | 435 | 91 | ||
More Information
Set Membership: |
|

jagn1
Genomic Map