Assay Details
| Gene Symbol: hist1h2aj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004821.1 | NP_001004821.1 | 1245 | 93 | ||
| GenBank mRNA | BC074601.1 | - | 1245 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hist1h2aj| Gene Symbol: hist1h2aj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004821.1 | NP_001004821.1 | 1245 | 93 | ||
| GenBank mRNA | BC074601.1 | - | 1245 | 93 | ||
Set Membership: |
|