Assay Details
| Gene Symbol: msrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004972.2 | NP_001004972.2 | 4 | 2748 | 64 | |
| GenBank mRNA | BC075488.1 | - | 4 | 2747 | 64 | |
| CX973900.1 | - | 1 | 123 | 64 | ||
More Information
Set Membership: |
|

msrb1
Genomic Map