Assay Details
| Gene Symbol: fzd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005669.1 | NP_001005669.1 | 1 | 1954 | 112 | |
| GenBank mRNA | BC074699.1 | - | 1 | 1954 | 112 | |
| CR926420.2 | - | 1 | 1826 | 112 | ||
More Information
Set Membership: |
|

fzd10
Genomic Map