Assay Details
| Gene Symbol: vkorc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006927.1 | NP_001006928.1 | 647 | 76 | ||
| GenBank mRNA | BC076993.1 | - | 647 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

vkorc1| Gene Symbol: vkorc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006927.1 | NP_001006928.1 | 647 | 76 | ||
| GenBank mRNA | BC076993.1 | - | 647 | 76 | ||
Set Membership: |
|