Assay Details
| Gene Symbol: tbx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007994.1 | NP_001007995.1 | 1343 | 67 | ||
| GenBank mRNA | BC080873.1 | - | 1343 | 67 | ||
| CR942588.2 | - | 1339 | 67 | |||
More Information
Set Membership: |
|

tbx6
Genomic Map