Assay Details
| Gene Symbol: ninj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008020.1 | NP_001008021.2 | 3 | 652 | 94 | |
| GenBank mRNA | BC080899.1 | - | 3 | 652 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ninj1| Gene Symbol: ninj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008020.1 | NP_001008021.2 | 3 | 652 | 94 | |
| GenBank mRNA | BC080899.1 | - | 3 | 652 | 94 | |
Set Membership: |
|