Assay Details
| Gene Symbol: prr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008030.2 | NP_001008031.2 | 1167 | 90 | ||
| GenBank mRNA | BC080912.1 | - | 1051 | 90 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

prr5| Gene Symbol: prr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008030.2 | NP_001008031.2 | 1167 | 90 | ||
| GenBank mRNA | BC080912.1 | - | 1051 | 90 | ||
Set Membership: |
|