Assay Details
| Gene Symbol: prss1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011251.1 | NP_001011251.1 | 5 | 950 | 94 | |
| Gene Symbol: LOC105946127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC087830.1 | - | 5 | 950 | 94 | |
More Information
Set Membership: |
|

prss1.2
Genomic Map