Assay Details
| Gene Symbol: nfil3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015710.1 | NP_001015710.1 | 1 | 1348 | 100 | |
| XM_012961583.2 | XP_012817037.1 | 2 | 1726 | 100 | ||
| XM_012961589.2 | XP_012817043.1 | 3 | 1773 | 100 | ||
| GenBank mRNA | BC089641.1 | - | 1 | 1348 | 100 | |
More Information
Set Membership: |
|

nfil3
Genomic Map