Assay Details
| Gene Symbol: ca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015729.1 | NP_001015729.1 | 7 | 1450 | 111 | |
| XM_012964640.2 | XP_012820094.1 | 8 | 1591 | 111 | ||
| GenBank mRNA | BC089661.1 | - | 7 | 1450 | 111 | |
| BC121554.1 | - | 7 | 1336 | 111 | ||
| BC160798.1 | - | 7 | 1388 | 121 | ||
More Information
Set Membership: |
|

ca2
Genomic Map