Assay Details
| Gene Symbol: park7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015851.1 | NP_001015851.1 | 6 | 1749 | 88 | |
| GenBank mRNA | BC090355.1 | - | 6 | 1749 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

park7| Gene Symbol: park7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015851.1 | NP_001015851.1 | 6 | 1749 | 88 | |
| GenBank mRNA | BC090355.1 | - | 6 | 1749 | 88 | |
Set Membership: |
|