Assay Details
| Gene Symbol: zfp160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016517.2 | NP_001016517.1 | 4 | 1037 | 126 | |
| XM_018089483.1 | XP_017944972.1 | 5 | 1152 | 126 | ||
| GenBank mRNA | CR926215.2 | - | 4 | 1037 | 126 | |
More Information
Set Membership: |
|

zfp160
Genomic Map