Assay Details
| Gene Symbol: ints9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016601.2 | NP_001016601.1 | 16 | 1754 | 68 | |
| GenBank mRNA | CR760554.2 | - | 16 | 1754 | 68 | |
| Gene Symbol: ints9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018089551.1 | XP_017945040.1 | 16 | 2070 | 68 | |
More Information
Set Membership: |
|

ints9
Genomic Map