Assay Details
| Gene Symbol: hist2h3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016636.3 | NP_001016636.1 | 1 | 153 | 64 | |
| GenBank mRNA | BC157346.1 | - | 140 | 64 | ||
| CR855729.2 | - | 1 | 153 | 64 | ||
| Gene Symbol: LOC100486113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012953875.2 | XP_012809329.1 | 1 | 238 | 64 | |
| Gene Symbol: LOC100493041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018092769.1 | XP_017948258.1 | 1 | 175 | 64 | |
| Gene Symbol: LOC100495338 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002942293.4 | XP_002942339.1 | 1 | 207 | 64 | |
| Gene Symbol: LOC100495788 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004916982.2 | XP_004917039.1 | 1 | 726 | 64 | |
| Gene Symbol: LOC100496129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018092772.1 | XP_017948261.1 | 1 | 171 | 64 | |
| Gene Symbol: LOC101731144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018092773.1 | XP_017948262.1 | 1 | 200 | 64 | |
| Gene Symbol: LOC108646286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018092771.1 | XP_017948260.1 | 1 | 178 | 64 | |
| Gene Symbol: LOC108646287 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018092774.1 | XP_017948263.1 | 1 | 241 | 64 | |
More Information
Set Membership: |
|

hist2h3c
Genomic Map