Assay Details
| Gene Symbol: tmem11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016689.2 | NP_001016689.1 | 2 | 446 | 113 | |
| GenBank mRNA | CR760191.2 | - | 2 | 446 | 113 | |
More Information
Set Membership: |
|

tmem11
Genomic Map