Assay Details
| Gene Symbol: gbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016773.2 | NP_001016773.1 | 7 | 1044 | 101 | |
| GenBank mRNA | CR855726.2 | - | 7 | 1044 | 101 | |
More Information
Set Membership: |
|

gbp1
Genomic Map