Assay Details
| Gene Symbol: aurkaip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016966.1 | NP_001016966.1 | 4 | 955 | 108 | |
| XM_012966643.2 | XP_012822097.1 | 4 | 1008 | 108 | ||
| GenBank mRNA | BC075331.1 | - | 4 | 990 | 108 | |
| CR759987.2 | - | 4 | 955 | 108 | ||
More Information
Set Membership: |
|

aurkaip1
Genomic Map