Assay Details
| Gene Symbol: pole3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017264.2 | NP_001017264.1 | 5 | 744 | 101 | |
| XM_012969763.2 | XP_012825217.1 | 5 | 809 | 101 | ||
| GenBank mRNA | BC135964.1 | - | 5 | 749 | 101 | |
| CR760000.2 | - | 5 | 744 | 101 | ||
More Information
Set Membership: |
|

pole3
Genomic Map