Assay Details
| Gene Symbol: dand5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044459.1 | NP_001037924.1 | 347 | 100 | ||
| NM_001044497.1 | NP_001037962.1 | 340 | 100 | |||
| GenBank mRNA | BC155373.1 | - | 308 | 100 | ||
| CR761079.2 | - | 340 | 100 | |||
| CR761214.2 | - | 347 | 100 | |||
More Information
Set Membership: |
|

dand5
Genomic Map