Assay Details
| Gene Symbol: cdk5r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078790.1 | NP_001072258.1 | 1 | 1049 | 81 | |
| GenBank mRNA | BC121244.1 | - | 1 | 1049 | 81 | |
| BC136097.1 | - | 1 | 1028 | 81 | ||
| CT030074.2 | - | 1 | 1060 | 81 | ||
More Information
Set Membership: |
|

cdk5r2
Genomic Map