Assay Details
| Gene Symbol: zfp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078916.1 | NP_001072384.1 | 4 | 1048 | 73 | |
| GenBank mRNA | BC121549.1 | - | 4 | 1048 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

zfp2| Gene Symbol: zfp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078916.1 | NP_001072384.1 | 4 | 1048 | 73 | |
| GenBank mRNA | BC121549.1 | - | 4 | 1048 | 73 | |
Set Membership: |
|