Assay Details
| Gene Symbol: znf519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102709.1 | NP_001096179.1 | 1351 | 103 | ||
| GenBank mRNA | BC133718.1 | - | 1351 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

znf519| Gene Symbol: znf519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102709.1 | NP_001096179.1 | 1351 | 103 | ||
| GenBank mRNA | BC133718.1 | - | 1351 | 103 | ||
Set Membership: |
|