Assay Details
| Gene Symbol: rabl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102825.1 | NP_001096295.1 | 1249 | 88 | ||
| GenBank mRNA | BC135594.1 | - | 1249 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rabl3| Gene Symbol: rabl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102825.1 | NP_001096295.1 | 1249 | 88 | ||
| GenBank mRNA | BC135594.1 | - | 1249 | 88 | ||
Set Membership: |
|