Assay Details
| Gene Symbol: timm10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102846.2 | NP_001096316.1 | 452 | 108 | ||
| GenBank mRNA | CX370796.2 | - | 454 | 108 | ||
| DN035373.1 | - | 404 | 108 | |||
| DT431530.1 | - | 582 | 108 | |||
More Information
Set Membership: |
|

timm10b
Genomic Map