Assay Details
Gene Symbol: nrarp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102879.1 | NP_001096349.1 | 446 | 85 | ||
GenBank mRNA | BC135819.1 | - | 446 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
